siRNA Details
| Gene Symbol: Bahcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198423.3 | NP_940815.3 | 24, 25 | 6501 | ||
| GenBank mRNA | AB257855.1 | - | 23, 24 | 6503 | ||
| AK147250.1 | - | 24, 25 | 6501 | |||
| AK147400.1 | - | 16, 17 | 3409 | |||
| AK164229.1 | - | 7, 8 | 742 | |||
| BC060615.1 | - | 7, 8 | 773 | |||
| BC072602.1 | - | 13, 14 | 1939 | 6830770 | ||

Bahcc1
Genomic Map