siRNA Details
| Gene Symbol: Greb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252071.1 | NP_001239000.1 | 30 | 5566 | ||
| NM_015764.4 | NP_056579.2 | 32 | 5972 | |||
| GenBank mRNA | AB093248.1 | - | 15 | 2900 | ||
| AF180470.1 | - | 31 | 5628 | |||
| AK017875.1 | - | 7 | 1189 | |||
| AK033573.1 | - | 2 | 131 | |||
| AK147337.1 | - | 30 | 5500 | |||
| BC160931.1 | - | 31 | 5498 | |||

Greb1
Genomic Map