siRNA Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024645.1 | NP_001019816.1 | 9, 10 | 838 | ||
| GenBank mRNA | AK131164.1 | - | 9, 10 | 781 | ||
| AK148355.1 | - | 9, 10 | 861 | |||
| AK158466.1 | - | 10, 11 | 1084 | |||
| BC082805.1 | - | 9, 10 | 758 | |||
| BC092223.1 | - | 9, 10 | 756 | |||

Lrrc16b
Genomic Map