siRNA Details
| Gene Symbol: Lrrc16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024645.1 | NP_001019816.1 | 11 | 985 | ||
| GenBank mRNA | AK131164.1 | - | 11 | 928 | ||
| AK148355.1 | - | 11 | 1008 | |||
| AK158466.1 | - | 12 | 1231 | |||
| BC082805.1 | - | 11 | 905 | |||
| BC092223.1 | - | 11 | 903 | |||

Lrrc16b
Genomic Map