siRNA Details
| Gene Symbol: Abcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011994.2 | NP_036124.2 | 10 | 2261 | ||
| GenBank mRNA | AK051445.1 | - | 10 | 2235 | ||
| AK082588.1 | - | 10 | 2233 | |||
| AK087653.1 | - | 10 | 1880 | |||
| AK134763.1 | - | 5 | 572 | |||
| AK161027.1 | - | 10 | 1922 | |||
| BC019187.1 | - | 10 | 2205 | 5027149 | ||
| Z48670.1 | - | 10 | 2265 | |||

Abcd2
Genomic Map