siRNA Details
| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175546.4 | NP_780755.2 | 11 | 1314 | ||
| GenBank mRNA | AK086275.1 | - | 11 | 1327 | ||
| AK131958.1 | - | 11 | 1313 | |||
| AK151817.1 | - | 11 | 1270 | |||
| BC116632.1 | - | 11 | 1260 | 40056912 | ||
| CB520148.1 | - | 2 | 39 | |||

Wdfy2
Genomic Map