siRNA Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178747.3 | NP_848862.1 | 5 | 532 | ||
| GenBank mRNA | AK077740.1 | - | 5 | 533 | ||
| AK167460.1 | - | 5 | 521 | |||
| AY453064.1 | - | 4 | 369 | |||
| BC019856.1 | - | 5 | 479 | 5123684 | ||
| BC028822.1 | - | 5 | 488 | 5097681 | ||
| BC028828.1 | - | 5 | 491 | 5101228 | ||
| BC034835.1 | - | 4 | 288 | |||

Gulo
Genomic Map