siRNA Details
| Gene Symbol: 9930012K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004155.2 | NP_001004155.2 | 2 | 567 | ||
| NM_001112735.1 | NP_001106206.1 | 2 | 567 | |||
| GenBank mRNA | AK132259.1 | - | 2 | 544 | ||
| BC032212.1 | - | 2 | 507 | |||
| BC080738.1 | - | 2 | 547 | 30433488 | ||

9930012K11Rik
Genomic Map