siRNA Details
| Gene Symbol: Casp8ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122978.1 | NP_001116450.1 | 7 | 1030 | ||
| NM_011997.2 | NP_036127.2 | 7 | 1277 | |||
| GenBank mRNA | AF132726.1 | - | 7 | 1237 | ||
| AK142148.1 | - | 7 | 1029 | |||
| AK154086.1 | - | 1 | 12 | |||
| AK165109.1 | - | 7 | 1027 | |||
| BC138022.1 | - | 6 | 1020 | 8861041 | ||
| BC138041.1 | - | 6 | 1020 | 8861060 | ||

Casp8ap2
Genomic Map