siRNA Details
| Gene Symbol: Lsamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175548.3 | NP_780757.1 | 5 | 736 | ||
| GenBank mRNA | AK030681.1 | - | 5 | 1042 | ||
| AK044845.1 | - | 5 | 736 | |||
| AK140043.1 | - | 5 | 873 | |||
| AK158702.1 | - | 5 | 1220 | |||
| AK164171.1 | - | 1043 | ||||

Lsamp
Genomic Map