siRNA Details
| Gene Symbol: Nrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173440.2 | NP_775616.1 | 4 | 2967 | ||
| GenBank mRNA | AF053062.1 | - | 2 | 2615 | ||
| AK036273.1 | - | 4 | 2968 | |||
| AK156233.1 | - | 4 | 2946 | |||
| AK157467.1 | - | 4 | 2948 | |||
| BC060232.1 | - | 3 | 2765 | 6852643 | ||
| BC075638.1 | - | 4 | 2844 | |||

Nrip1
Genomic Map