siRNA Details
| Gene Symbol: Capn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013767.2 | NP_001013789.1 | 19 | 1952 | ||
| GenBank mRNA | AY578330.1 | - | 19 | 1952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn11| Gene Symbol: Capn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013767.2 | NP_001013789.1 | 19 | 1952 | ||
| GenBank mRNA | AY578330.1 | - | 19 | 1952 | ||