siRNA Details
| Gene Symbol: Med14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048208.1 | NP_001041673.1 | 17 | 2461 | ||
| NM_012005.3 | NP_036135.3 | 17 | 2461 | |||
| GenBank mRNA | AK143808.1 | - | 3 | 403 | ||
| BC065072.1 | - | 17 | 2561 | 6830820 | ||

Med14
Genomic Map