siRNA Details
| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172964.4 | NP_766552.3 | 14, 15 | 1948 | ||
| GenBank mRNA | AK087523.1 | - | 14, 15 | 1961 | ||
| AK133720.1 | - | 14, 15 | 1787 | |||
| AK141901.1 | - | 14, 15 | 1898 | |||
| AK162051.1 | - | 8, 9 | 988 | |||
| AK173147.1 | - | 13, 14 | 1575 | |||
| BC066788.1 | - | 14, 15 | 1938 | 30049271 | ||

Arhgap28
Genomic Map