siRNA Details
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011500.2 | NP_035630.2 | 10 | 1290 | ||
| GenBank mRNA | AJ223777.1 | - | 10 | 1217 | ||
| AK135684.1 | - | 5 | 482 | |||
| AK136032.1 | - | 8 | 939 | |||
| BC090968.1 | - | 10 | 1290 | 30646517 | ||
| BC150727.1 | - | 10 | 1258 | 9087638 | ||

Strn
Genomic Map