siRNA Details
| Gene Symbol: Sap130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172965.2 | NP_766553.1 | 16 | 2476 | ||
| GenBank mRNA | AK032704.1 | - | 6 | 1141 | ||
| AK034401.1 | - | 16 | 2461 | |||
| AK053675.1 | - | 16 | 2477 | |||
| BC036975.1 | - | 6 | 1167 | |||
| BC063075.1 | - | 13 | 2003 | 5721433 | ||
| BC066030.1 | - | 16 | 2491 | |||

Sap130
Genomic Map