siRNA Details
| Gene Symbol: Stk32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178749.3 | NP_848864.1 | 12 | 1298 | ||
| GenBank mRNA | AK036266.1 | - | 12 | 1299 | ||
| AK042599.1 | - | 12 | 1412 | |||
| AK044474.1 | - | 12 | 1308 | |||
| BC055002.1 | - | 1228 | 6313304 | |||

Stk32a
Genomic Map