siRNA Details
| Gene Symbol: Dysf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077694.2 | NP_001071162.1 | 43 | 5109 | ||
| NM_021469.3 | NP_067444.2 | 43 | 4927 | |||
| GenBank mRNA | AF188290.2 | - | 42 | 4702 | ||
| AK087986.1 | - | 7 | 799 | |||
| AK131144.1 | - | 43 | 4924 | |||
| BC043692.1 | - | 3 | 299 | |||

Dysf
Genomic Map