siRNA Details
| Gene Symbol: Dysf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077694.2 | NP_001071162.1 | 35, 36 | 4255 | ||
| NM_021469.3 | NP_067444.2 | 35, 36 | 4073 | |||
| GenBank mRNA | AF188290.2 | - | 34, 35 | 3848 | ||
| AK131144.1 | - | 35, 36 | 4070 | |||

Dysf
Genomic Map