siRNA Details
| Gene Symbol: Cpsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164272.1 | NP_001157744.1 | 3 | 350 | ||
| NM_172302.3 | NP_758506.3 | 3 | 350 | |||
| GenBank mRNA | AK044306.1 | - | 3 | 329 | ||
| AK053375.1 | - | 3 | 329 | |||
| AK088603.1 | - | 3 | 328 | |||
| AK165330.1 | - | 3 | 328 | |||
| AK188327.1 | - | 2 | 218 | |||
| AK189206.1 | - | 2 | 218 | |||
| AK196685.1 | - | 237 | ||||
| BC017519.1 | - | 2 | 270 | |||
| BC038812.1 | - | 2 | 280 | 4506179 | ||

Cpsf7
Genomic Map