siRNA Details
| Gene Symbol: Exo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012012.4 | NP_036142.2 | 12 | 1839 | ||
| GenBank mRNA | AK028728.1 | - | 12 | 1820 | ||
| BC006671.1 | - | 12 | 1803 | 3499406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exo1| Gene Symbol: Exo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012012.4 | NP_036142.2 | 12 | 1839 | ||
| GenBank mRNA | AK028728.1 | - | 12 | 1820 | ||
| BC006671.1 | - | 12 | 1803 | 3499406 | ||