siRNA Details
| Gene Symbol: Exo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012012.4 | NP_036142.2 | 12 | 2226 | ||
| GenBank mRNA | AJ238213.1 | - | 10 | 2081 | ||
| AK028728.1 | - | 12 | 2207 | |||
| AK166425.1 | - | 12 | 2202 | |||
| BC006671.1 | - | 12 | 2190 | 3499406 | ||

Exo1
Genomic Map