siRNA Details
| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199021.3 | NP_950186.3 | 13 | 1270 | ||
| GenBank mRNA | AK032579.1 | - | 11 | 916 | ||
| AK046842.1 | - | 13 | 1271 | |||
| BC063074.1 | - | 3 | 192 | 5697531 | ||
| BC067026.1 | - | 13 | 1263 | 6853947 | ||

Dpp10
Genomic Map