siRNA Details
| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199021.3 | NP_950186.3 | 2 | 255 | ||
| GenBank mRNA | AK046842.1 | - | 2 | 257 | ||
| BC067026.1 | - | 2 | 248 | 6853947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpp10| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199021.3 | NP_950186.3 | 2 | 255 | ||
| GenBank mRNA | AK046842.1 | - | 2 | 257 | ||
| BC067026.1 | - | 2 | 248 | 6853947 | ||