siRNA Details
| Gene Symbol: Colgalt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177756.4 | NP_808424.3 | 12 | 1974 | ||
| GenBank mRNA | AK052838.1 | - | 12 | 1975 | ||
| AK076222.1 | - | 12 | 1718 | |||
| AK085716.1 | - | 14 | 2166 | |||
| AK134084.1 | - | 12 | 1955 | |||
| BC068118.1 | - | 12 | 1920 | 5706069 | ||

Colgalt2
Genomic Map