siRNA Details
| Gene Symbol: Inpp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290797.1 | NP_001277726.1 | 12 | 1420 | ||
| NM_001290798.1 | NP_001277727.1 | 2 | 296 | |||
| NM_001290799.1 | NP_001277728.1 | 2 | 296 | |||
| NM_030266.4 | NP_084542.2 | 12 | 1420 | |||
| NM_172971.3 | NP_766559.2 | 2 | 296 | |||
| GenBank mRNA | AF317838.1 | - | 10 | 973 | ||
| AK035867.1 | - | 2 | 297 | |||
| AK149414.1 | - | 12 | 1345 | |||
| BC120541.1 | - | 2 | 292 | 40129464 | ||
| BC144747.1 | - | 2 | 292 | |||
| BC144748.1 | - | 2 | 292 | 9053296 | ||

Inpp4a
Genomic Map