siRNA Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290801.1 | NP_001277730.1 | 12, 13 | 1648 | ||
| NM_173870.3 | NP_776295.1 | 13, 14 | 1727 | |||
| GenBank mRNA | AB053217.1 | - | 12, 13 | 1403 | ||
| AK052278.1 | - | 10, 11 | 1224 | |||
| AK155724.1 | - | 13, 14 | 1729 | |||
| AK163327.1 | - | 12, 13 | 1480 | |||

Mgat4a
Genomic Map