siRNA Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012017.2 | NP_036147.1 | 5 | 634 | ||
| GenBank mRNA | AF083339.1 | - | 5 | 632 | ||
| AK185041.1 | - | 3 | 300 | |||
| BB612150.1 | - | 4 | 491 | |||
| BC055075.1 | - | 5 | 628 | 30056643 | ||
| CV562260.1 | - | 4 | 430 | |||

Zfp346
Genomic Map