siRNA Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012017.2 | NP_036147.1 | 1, 2 | 212 | ||
| GenBank mRNA | AF083339.1 | - | 1, 2 | 210 | ||
| AK178507.1 | - | 1, 2 | 20 | |||
| AK197315.1 | - | 1, 2 | 20 | |||
| BB612150.1 | - | 1, 2 | 214 | |||
| BC055075.1 | - | 1, 2 | 206 | 30056643 | ||

Zfp346
Genomic Map