siRNA Details
| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175031.3 | NP_778196.2 | 21 | 2669 | ||
| GenBank mRNA | AK029842.1 | - | 21 | 2579 | ||
| AK047284.1 | - | 10 | 1122 | |||
| AK086716.1 | - | 5 | 457 | |||
| AK173142.1 | - | 20 | 2535 | |||
| BC043103.1 | - | 21 | 2621 | 6406485 | ||
| BC058698.1 | - | 18 | 2350 | 6831702 | ||
| BC061470.1 | - | 20 | 2478 | |||

Stk36
Genomic Map