siRNA Details
| Gene Symbol: BC035947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032005.1 | - | 2 | 1643 | ||
| BC138135.1 | - | 2 | 1597 | 8861154 | ||
| BC138136.1 | - | 2 | 1597 | 8861155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC035947| Gene Symbol: BC035947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032005.1 | - | 2 | 1643 | ||
| BC138135.1 | - | 2 | 1597 | 8861154 | ||
| BC138136.1 | - | 2 | 1597 | 8861155 | ||