siRNA Details
| Gene Symbol: Polr3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164096.1 | NP_001157568.1 | 5 | 451 | ||
| NM_025298.3 | NP_079574.2 | 6 | 529 | |||
| GenBank mRNA | AK005034.1 | - | 5 | 452 | ||
| AK012184.1 | - | 6 | 489 | |||
| AK144721.1 | - | 6 | 491 | |||
| AK146279.1 | - | 6 | 486 | |||
| AK159723.1 | - | 6 | 486 | |||
| AK218372.1 | - | 2 | 89 | |||
| BC028924.1 | - | 6 | 455 | |||
| BC037637.1 | - | 6 | 479 | 5010681 | ||

Polr3e
Genomic Map