siRNA Details
| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253897.1 | NP_001240826.1 | 4 | 477 | ||
| NM_001253898.1 | NP_001240827.1 | 3 | 299 | |||
| NM_012029.2 | NP_036159.1 | 4 | 485 | |||
| GenBank mRNA | AF182510.1 | - | 3 | 498 | ||
| AK007564.1 | - | 4 | 459 | |||
| AK012835.1 | - | 4 | 457 | |||
| AK219052.1 | - | 1 | 23 | |||
| BC004583.1 | - | 3 | 286 | 3587327 | ||
| BC016424.1 | - | 4 | 425 | 4924678 | ||
| BY123162.1 | - | 3 | 279 | |||
| CB589832.1 | - | 2 | 181 | |||

Ecsit
Genomic Map