siRNA Details
| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253897.1 | NP_001240826.1 | 7, 8 | 1216 | ||
| NM_001253898.1 | NP_001240827.1 | 6, 7 | 1038 | |||
| NM_012029.2 | NP_036159.1 | 7, 8 | 1224 | |||
| GenBank mRNA | AF182510.1 | - | 6, 7 | 1237 | ||
| AK007564.1 | - | 7, 8 | 1190 | |||
| AK012835.1 | - | 7, 8 | 1189 | |||
| BC004583.1 | - | 6, 7 | 1025 | 3587327 | ||
| BC016424.1 | - | 7, 8 | 1164 | 4924678 | ||

Ecsit
Genomic Map