siRNA Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012031.3 | NP_036161.2 | 10 | 1208 | ||
| GenBank mRNA | AF181252.1 | - | 10 | 1200 | ||
| AK032601.1 | - | 7 | 1003 | |||
| BC046313.1 | - | 10 | 1208 | 6310604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag1| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012031.3 | NP_036161.2 | 10 | 1208 | ||
| GenBank mRNA | AF181252.1 | - | 10 | 1200 | ||
| AK032601.1 | - | 7 | 1003 | |||
| BC046313.1 | - | 10 | 1208 | 6310604 | ||