siRNA Details
| Gene Symbol: Plch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177732.1 | NP_001171203.1 | 24 | 4439 | ||
| NM_001177733.1 | NP_001171204.1 | 24 | 4434 | |||
| NM_183191.3 | NP_899014.2 | 23 | 4411 | |||
| GenBank mRNA | AK173095.1 | - | 14 | 3234 | ||
| AY691172.1 | - | 24 | 4439 | |||
| AY691173.1 | - | 24 | 4434 | |||
| AY691174.1 | - | 23 | 4411 | |||
| BC042549.1 | - | 11 | 2703 | |||
| BC052372.1 | - | 11 | 2703 | |||
| BC055005.1 | - | 22 | 4425 | 6491217 | ||
| BC057610.1 | - | 3 | 2001 | |||

Plch1
Genomic Map