siRNA Details
| Gene Symbol: Tinag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012033.3 | NP_036163.3 | 1, 2 | 443 | ||
| GenBank mRNA | AF153366.1 | - | 1, 2 | 385 | ||
| AK008374.1 | - | 1, 2 | 411 | |||
| BC010745.1 | - | 1, 2 | 388 | 4239192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tinag| Gene Symbol: Tinag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012033.3 | NP_036163.3 | 1, 2 | 443 | ||
| GenBank mRNA | AF153366.1 | - | 1, 2 | 385 | ||
| AK008374.1 | - | 1, 2 | 411 | |||
| BC010745.1 | - | 1, 2 | 388 | 4239192 | ||