siRNA Details
| Gene Symbol: Wdr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175552.4 | NP_780761.1 | 11, 12 | 1421 | ||
| GenBank mRNA | AK043557.1 | - | 11, 12 | 1354 | ||
| AK050738.1 | - | 11, 12 | 1389 | |||
| AK050797.1 | - | 11, 12 | 1392 | |||
| BC050990.1 | - | 2, 3 | 350 | |||
| BC063100.1 | - | 11, 12 | 1407 | 6332749 | ||

Wdr3
Genomic Map