siRNA Details
| Gene Symbol: Fbxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172988.4 | NP_766576.1 | 9 | 2151 | ||
| GenBank mRNA | AI836810.1 | - | 1 | 409 | ||
| AK028786.1 | - | 9 | 3274 | |||
| AK032391.1 | - | 9 | 2130 | |||
| AK051429.1 | - | 9 | 2102 | |||
| AK083661.1 | - | 9 | 2088 | |||
| BC033265.1 | - | 10 | 2191 | 3983942 | ||

Fbxl4
Genomic Map