siRNA Details
| Gene Symbol: Cul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012042.3 | NP_036172.1 | 15 | 1966 | ||
| GenBank mRNA | AF083216.1 | - | 14 | 1690 | ||
| AF136441.1 | - | 14 | 1821 | |||
| AF176910.1 | - | 15 | 1855 | |||
| AK147004.1 | - | 15 | 1948 | |||
| AK164274.1 | - | 15 | 1933 | |||
| AK165978.1 | - | 15 | 1907 | |||
| BC004836.1 | - | 14 | 1775 | |||
| BC029260.1 | - | 15 | 1966 | 2812400 | ||

Cul1
Genomic Map