siRNA Details
| Gene Symbol: Golga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008146.3 | NP_032172.3 | 5 | 843 | ||
| GenBank mRNA | AK133623.1 | - | 5 | 782 | ||
| BC053002.1 | - | 5 | 843 | 5709074 | ||
| D78270.2 | - | 5 | 649 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golga3| Gene Symbol: Golga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008146.3 | NP_032172.3 | 5 | 843 | ||
| GenBank mRNA | AK133623.1 | - | 5 | 782 | ||
| BC053002.1 | - | 5 | 843 | 5709074 | ||
| D78270.2 | - | 5 | 649 | |||