siRNA Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040106.2 | NP_001035195.1 | 11 | 1779 | ||
| NM_177762.6 | NP_808430.2 | 11 | 1779 | |||
| GenBank mRNA | AK147363.1 | - | 11 | 1780 | ||
| BC141176.1 | - | 11 | 1535 | 9055788 | ||
| CF723295.1 | - | 4 | 299 | |||

Aak1
Genomic Map