siRNA Details
| Gene Symbol: Zfp384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252083.1 | NP_001239012.1 | 3, 4 | 386 | ||
| NM_175557.5 | NP_780766.3 | 3, 4 | 357 | |||
| GenBank mRNA | AK086185.1 | - | 3, 4 | 282 | ||
| AK156026.1 | - | 3, 4 | 388 | |||
| AK172476.1 | - | 3, 4 | 349 | |||
| BB662626.1 | - | 3, 4 | 282 | |||
| BC064013.1 | - | 3, 4 | 221 | 6515698 | ||

Zfp384
Genomic Map