siRNA Details
| Gene Symbol: Zfp384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252083.1 | NP_001239012.1 | 4 | 472 | ||
| NM_175557.5 | NP_780766.3 | 4 | 443 | |||
| GenBank mRNA | AK086185.1 | - | 4 | 368 | ||
| AK156026.1 | - | 4 | 474 | |||
| AK172476.1 | - | 4 | 435 | |||
| BB662626.1 | - | 4 | 368 | |||
| BC064013.1 | - | 4 | 307 | 6515698 | ||

Zfp384
Genomic Map