siRNA Details
| Gene Symbol: Zfp446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168561.1 | NP_001162033.1 | 5 | 1170 | ||
| NM_001168562.1 | NP_001162034.1 | 5 | 1586 | |||
| NM_175558.4 | NP_780767.1 | 6 | 1289 | |||
| GenBank mRNA | AK041761.1 | - | 6 | 1291 | ||
| AK138378.1 | - | 4 | 1588 | |||
| AK142766.1 | - | 5 | 1785 | |||
| BC048909.1 | - | 6 | 1231 | |||
| BC117850.1 | - | 5 | 1593 | |||
| BC117851.1 | - | 5 | 1172 | 40093910 | ||

Zfp446
Genomic Map