siRNA Details
Gene Symbol: Zfp446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168561.1 | NP_001162033.1 | 5 | 1325 | ||
NM_001168562.1 | NP_001162034.1 | 5 | 1741 | |||
NM_175558.4 | NP_780767.1 | 6 | 1444 | |||
GenBank mRNA | AK041761.1 | - | 6 | 1446 | ||
AK138378.1 | - | 4 | 1743 | |||
AK142766.1 | - | 5 | 1940 | |||
BC048909.1 | - | 6 | 1386 | |||
BC117850.1 | - | 5 | 1748 | |||
BC117851.1 | - | 5 | 1327 | 40093910 |
