siRNA Details
| Gene Symbol: Eif4e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039169.1 | NP_001034258.1 | 3 | 768 | ||
| NM_001039170.1 | NP_001034259.1 | 3 | 317 | |||
| NM_023314.3 | NP_075803.2 | 3 | 317 | |||
| GenBank mRNA | AF068116.1 | - | 3 | 231 | ||
| AK017221.1 | - | 3 | 769 | |||
| AK018516.1 | - | 3 | 315 | |||
| AK032965.1 | - | 3 | 321 | |||
| AK076067.1 | - | 1 | 32 | |||
| AK138055.1 | - | 3 | 256 | |||
| AK150495.1 | - | 3 | 256 | |||
| AK199854.1 | - | 3 | 299 | |||
| BC028903.1 | - | 3 | 367 | 4166275 | ||
| BC045153.1 | - | 3 | 251 | 6441327 | ||
| BC049077.1 | - | 3 | 285 | 5708556 | ||
| U01137.1 | - | 3 | 264 | |||

Eif4e2
Genomic Map