siRNA Details
| Gene Symbol: Eif4e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039169.1 | NP_001034258.1 | 6 | 1123 | ||
| NM_001039170.1 | NP_001034259.1 | 6 | 672 | |||
| NM_023314.3 | NP_075803.2 | 6 | 672 | |||
| GenBank mRNA | AK017221.1 | - | 6 | 1124 | ||
| AK018516.1 | - | 6 | 670 | |||
| AK032965.1 | - | 6 | 676 | |||
| AK076067.1 | - | 4 | 387 | |||
| AK138055.1 | - | 6 | 611 | |||
| AK150495.1 | - | 6 | 611 | |||
| BC028903.1 | - | 6 | 722 | 4166275 | ||
| BC045153.1 | - | 6 | 606 | 6441327 | ||
| BC049077.1 | - | 6 | 640 | 5708556 | ||
| U01137.1 | - | 6 | 620 | |||

Eif4e2
Genomic Map