siRNA Details
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012047.2 | NP_036177.1 | 12 | 1594 | ||
GenBank mRNA | AF084259.1 | - | 12 | 1567 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012047.2 | NP_036177.1 | 12 | 1594 | ||
GenBank mRNA | AF084259.1 | - | 12 | 1567 |