siRNA Details
| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012047.2 | NP_036177.1 | 11, 12 | 1565 | ||
| GenBank mRNA | AF084259.1 | - | 11, 12 | 1538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brd7| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012047.2 | NP_036177.1 | 11, 12 | 1565 | ||
| GenBank mRNA | AF084259.1 | - | 11, 12 | 1538 | ||